Merck
  • Inicio
  • Resultados de la búsqueda
  • Radical-scavenging Activity and Antioxidative Effects of Olive Leaf Components Oleuropein and Hydroxytyrosol in Comparison with Homovanillic Alcohol.
Radical-scavenging Activity and Antioxidative Effects of Olive Leaf Components Oleuropein and Hydroxytyrosol in Comparison with Homovanillic Alcohol.
Journal of oleo science (2015-07-03)
Aya Umeno, Mizuki Takashima, Kazutoshi Murotomi, Yoshihiro Nakajima, Taisuke Koike, Toshiki Matsuo, Yasukazu Yoshida
RESUMEN
Olive leaf has great potential as a natural antioxidant, and one of its major phenolic components is oleuropein. In this study, the antioxidant activity of oleuropein against oxygen-centered radicals was measured by examining its sparing effects on the peroxyl radical-induced decay of fluorescein and pyrogallol red, in comparison with related compounds. The antioxidant capacity of oleuropein against lipid peroxidation was also assessed through its effect on the free radical-induced oxidation of methyl linoleate in a micelle system. On a molar basis, oleuropein and hydroxytyrosol inhibited the decay of fluorescein for longer than both homovanillic alcohol and the vitamin-E mimic 2-carboxy-2,5,7,8-tetramethyl-6-chromanol (Trolox), but did not suppress pyrogallol red decay in a concentration-dependent manner. Measurement of the fluorescein decay period revealed that the stoichiometric number of oleuropein and hydroxytyrosol against peroxyl radicals was twice that of Trolox, which is substantially higher than expectations based on chemical structure. Oleuropein and hydroxytyrosol were also more effective than Trolox and homovanillic alcohol at suppressing the oxidation of methyl linoleate in the micelle system. Thus, both oleuropein and hydroxytyrosol exhibit high antioxidative activity against lipid peroxidation induced by oxygen-centered radicals, but the high reactivity of phenolic/catecholic radicals makes their mechanism of action complex.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Pyrogallol Red
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
SAFC
L-Cysteine
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%