  • Search Results
  • Modeling and Understanding Combination pMDI Formulations with Both Dissolved and Suspended Drugs.
Modeling and Understanding Combination pMDI Formulations with Both Dissolved and Suspended Drugs.
Molecular pharmaceutics (2015-08-11)
Stephen W Stein, Poonam Sheth, Usir S Younis, Erik Mogalian, Paul B Myrdal
ABSTRACT
A simulation model has been established to predict the residual aerodynamic particle size distribution (APSD) of dual-component pressurized metered dose inhalers (pMDIs). More specifically, this model estimates the APSD of pMDI formulations containing dissolved and suspended compounds for various formulations, and has been verified experimentally. Simulated and experimental data illustrate that APSDs of the dissolved and suspended components of the pMDI are influenced by concentrations of the dissolved and micronized suspended drugs, along with suspended drug size. Atomized droplets from such combination formulations may contain varying number of suspended drug particles and a representative concentration of dissolved drug. These sub-populations of atomized droplets may explain the residual APSDs. The suspended drug follows a monomodal, lognormal distribution and is more greatly impacted by the size and concentration of the suspended drug in comparison to the concentration of dissolved drug. On the other hand, dissolved drug illustrates a bimodal, lognormal residual particle size distribution both theoretically and experimentally. The smaller mode consists of residual particles made of dissolved drug only, while the larger mode consists of residual particles that contain both dissolved and suspended drugs. The model effectively predicted the size distributions of both the dissolved and suspended components of combination formulations (r(2) value of 0.914 for the comparison of simulated versus experimental MMAD values for the formulations examined). The results demonstrate that this model is a useful tool that may be able to expedite the development of combination pMDI formulation.
Sigma-Aldrich
Salbutamol hemisulfate salt, ≥98%
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Sigma-Aldrich
D-Lactose monohydrate, BioUltra, ≥99.5% (HPLC)
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.