  • Search Results
  • Synthesis, Characterization and Application of Cobalt Incorporated Mesoporous KIT-6 for the Visible Light Assisted Degradation of Methylene Blue.
Synthesis, Characterization and Application of Cobalt Incorporated Mesoporous KIT-6 for the Visible Light Assisted Degradation of Methylene Blue.
Journal of nanoscience and nanotechnology (2015-12-31)
N S Sanjini, B Winston, S Velmathi
ABSTRACT
Highly ordered mesoporous three dimensional la3d cobalt doped KIT-6 (Co-KIT-6) material was synthesized by the simple impregnation method and characterized by various physicochemical techniques such as X-ray diffraction, N2 adsorption-desorption isotherm, diffuse reflectance spectroscopy, Fourier transform infra-red spectroscopy and scanning electron microscopy. Larger pore diameter tuned with a narrow size distribution was observed for prepared Co-KIT-6. A series of Co-KIT-6 with different weight percentages solutions were prepared and the photocatalytic efficacy of the samples for degradation of methylene blue was studied under UV and visible light using UV-visible spectrophotometer. It was found that the catalytic activity depends on the amount of cobalt incorporated into the material. The complete degradation of dye was observed within 50 min. Different amount of catalyst and reaction time was considered for the optimization studies.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, puriss. p.a., ACS reagent, Ni ≤0.001%, ≥99.0% (KT)
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, reagent grade, 98%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG