  • Search Results
  • Initial studies on the direct and modulatory effects of nitric oxide on an identified central Helix aspersa neuron.
Initial studies on the direct and modulatory effects of nitric oxide on an identified central Helix aspersa neuron.
Invertebrate neuroscience : IN (2014-11-09)
Nicholas J D Wright, Lynda J Sides, Kerry Walling
ABSTRACT
The generation of the novel messenger molecule nitric oxide (NO) has been demonstrated in many tissues across phyla including nervous systems. It is produced on demand by the enzyme nitric oxide synthase often stimulated by intracellular calcium and typically affecting guanylate cyclase thought to be its principal target in an auto and/or paracrine fashion. This results in the generation of the secondary messenger cyclic guanosine monophosphate (cGMP). Nitric oxide synthase has been demonstrated in various mollusk brains and manipulation of NO levels has been shown to affect behavior in mollusks. Apart from modulation of the effect of the peptide GSPYFVamide, there appears little published on direct or modulatory effects of NO on Helix aspersa central neurons. We present here initial results to show that NO can be generated in the region around F1 in the right parietal ganglion and that NO and cGMP directly hyperpolarize this neuron. For example, application of the NO-donor S-nitroso-N-acetyl-D,L-penicillamine (SNAP; 200 µM) can cause a mean hyperpolarization of 41.7 mV, while 2 mM 8-bromo-cyclic guanosine monophosphate (8-bromo-cGMP) produced a mean hyperpolarization of 33.4 mV. Additionally, pre-exposure to NO-donors or cGMP appears to significantly reduce or even eliminates the normal hyperpolarizing K(+)-mediated response to dopamine (DA) by this neuron; 200 µM SNAP abolishes a standard response to 0.5 µM DA while 1 mM 8-bromo-cGMP reduces it 62%.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Methylene Blue solution, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene blue, certified by the Biological Stain Commission
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.5% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Calcium, granular, 99%