  • Search Results
  • Antinociceptive Activity and Toxicity Evaluation of the Fatty Oil from Plukenetia polyadenia Mull. Arg. (Euphorbiaceae).
Antinociceptive Activity and Toxicity Evaluation of the Fatty Oil from Plukenetia polyadenia Mull. Arg. (Euphorbiaceae).
Molecules (Basel, Switzerland) (2015-05-06)
Amanda S Mota, Anderson B de Lima, Thayana Lucy F Albuquerque, Tiago S Silveira, José Luiz M do Nascimento, Joyce Kelly R da Silva, Alcy F Ribeiro, José Guilherme S Maia, Gilmara N T Bastos
ABSTRACT
Seed oil (Pp-oil) of Plukenetia polyadenia is used by native people of the Brazilian Amazon against arthritis and rheumatism, spreading it on the arms and legs to reduce the pain and inflammation. Pp-oil was obtained by pressing dried seeds at room temperature to give a 47.0% yield of oil. It was then subjected to fatty acid composition analysis. The principal fatty acids were linoleic acid (46.5%), α-linolenic acid (34.4%) and oleic acid (13.9%). Then, it was evaluated for its antinociceptive activity in mice, using the acetic acid-induced abdominal writhing, hot plate and formalin test models. Additionally, its toxicity was determined. The Pp-oil proved to have no toxicological effects, showing dose-dependent antinociceptive effect under chemical stimulation. At oral doses of 25-100 mg/kg, Pp-oil significantly reduced the abdominal writhes in the writhing test. A higher oral dose of 200 mg/kg did not induce alterations in the latency time of the hot plate test when compared to the control, suggesting an analgesic activity of peripheral origin. At oral doses of 50 and 100 mg/kg, the Pp-oil significantly reduced the second phase of the algic stimulus in the formalin test. In addition, the antinociception of Pp-oil was reversed by naloxone in the evaluation of its mechanism of action. Therefore, the Pp-oil proved to be safe at very high doses and to show significant analgesic properties. The role of Pp-oil is still being investigated with respect the mechanism of action, but the results suggest that opiod receptors could be involved in the antinociception action observed for the oil of P. polyadenia.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Sodium methoxide, reagent grade, 95%, powder
Sigma-Aldrich
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)
Sigma-Aldrich
Sodium methoxide solution, 25 wt. % in methanol
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Diethyl ether