  • Search Results
  • Development of a high-throughput in vitro intestinal lipolysis model for rapid screening of lipid-based drug delivery systems.
Development of a high-throughput in vitro intestinal lipolysis model for rapid screening of lipid-based drug delivery systems.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-07-15)
Mette D Mosgaard, Philip Sassene, Huiling Mu, Thomas Rades, Anette Müllertz
ABSTRACT
To develop a high-throughput in vitro intestinal lipolysis (HTP) model, without any means of pH-stat-titration, to enable a fast evaluation of lipid-based drug delivery systems (LbDDS). The HTP model was compared to the traditionally used dynamic in vitro lipolysis (DIVL) model with regard to the extent of lipid digestion and drug distribution of two poorly soluble model drugs (cinnarizine and danazol), during digestion of three LbDDS (LbDDS I-III). The HTP model was able to maintain pH around 6.5 during digestion, without the addition of NaOH to neutralize the free fatty acids (FFAs), due to an increased buffer capacity. Cinnarizine was primarily located in the aqueous phase during digestion of all three LbDDS and did not differ significantly between the two models. The distribution of danazol varied from formulation to formulation, but no significant difference between the models was observed. The triacylglycerides (TAG) in LbDDS III were digested to the same extent in both models, whereas the TAG present in LbDDS II was digested slightly less in the HTP model. No TAG was present in LbDDS I and digestion was therefore not analyzed. The HTP model is able to predict drug distribution during digestion of LbDDS containing poorly water soluble drugs in the same manner as the DIVL model. Thus the HTP model might prove applicable for high-throughput evaluation of LbDDS in e.g. 96 well plates or small scale dissolution equipment.
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
SAFC
BIS-TRIS
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
SAFC
BIS-TRIS
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Pancreatin from porcine pancreas, ≥3 × USP specifications
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Sesame oil, Antioxidant, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
4-Bromophenylboronic acid, ≥95.0%
Sigma-Aldrich
Danazol, ≥98%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Cinnarizine, powder
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O