• Inicio
  • Resultados de la búsqueda
  • Photosensitizer and Light Pave the Way for Cytosolic Targeting and Generation of Cytosolic CD8 T Cells Using PLGA Vaccine Particles.
Photosensitizer and Light Pave the Way for Cytosolic Targeting and Generation of Cytosolic CD8 T Cells Using PLGA Vaccine Particles.
Journal of immunology (Baltimore, Md. : 1950) (2015-05-29)
Cristina Bruno, Ying Waeckerle-Men, Monika Håkerud, Thomas M Kündig, Bruno Gander, Pål Johansen
RESUMEN
The generation of CTLs is crucial in the immunological fight against cancer and many infectious diseases. To achieve this, vaccine Ags need to be targeted to the cytosol of dendritic cells, which can activate CD8 T cells via MHC class I (MHCI). Therefore, such targeting has become one of the major objectives of vaccine research. In this study, we aimed to bypass the unwanted and default MHC class II Ag presentation and trigger MHCI presentation by using a photosensitizer that, upon light activation, would facilitate cytosolic targeting of codelivered Ag. Poly(lactide-co-glycolide) microparticles ∼1 μm size were loaded with OVA and the photosensitizer tetraphenyl chlorine disulphonate (TPCS2a) and administered intradermally in mice, which were illuminated 1 d later for activation of the photosensitizer. Immunization in the presence of TPCS2a significantly increased activation of CD8 T cells compared with immunization without TPCS2a and as measured by CD8 T cell proliferation, production of proinflammatory IFN-γ, TNF-α, and IL-2, and prevention of tumor growth. Cytotoxicity was demonstrated by granzyme B production in vitro and by in vivo killing of CFSE-labeled targets. CD4-dependent Ab responses were abrogated in mice immunized with TPCS2a-containing particles, suggesting that photosensitization facilitated a shift from default MHC class II toward MHCI Ag presentation. Hence, vaccine particles with Ag and photosensitizers proved an effective vehicle or adjuvant for stimulation of CTLs, and they may find potential application in therapeutic cancer vaccination and in prophylactic and therapeutic vaccination against intracellular infections.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Chlorhexidine diacetate salt hydrate, bis(biguanide) antimicrobial
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®