Heterogeneous distribution of axonal cytoskeleton proteins in the human optic nerve.
Investigative ophthalmology & visual science (2009-01-27)
Chandrakumar Balaratnasingam, William H Morgan, Victoria Johnstone, Stephen J Cringle, Dao-Yi Yu
RESUMEN
Cytoskeleton proteins play a critical role in maintaining retinal ganglion cell structure, viability, and function. This study documents the distribution of cytoskeleton protein subunits in the various regions of the normal human optic nerve and identifies important relationships among mitochondria, myelin, and neurofilament proteins. Twenty-three optic nerves from human cadavers were used. Confocal microscopy was used to examine the distribution of neurofilament light, neurofilament medium, neurofilament heavy (phosphorylated and unphosphorylated), neurofilament heavy (phosphorylated only), actin, and microtubule associated protein (MAP)-1 along the sagittal plane of the optic nerve. Comparisons were made among superior, middle, and inferior regions and also among temporal, central, and nasal portions of the optic nerve. Colocalization of neurofilament light, mitochondrial cytochrome c oxidase (COX), and myelin was also performed. There are significant differences in the pattern and distribution of neurofilament protein subunits, actin, and MAP-1 along the sagittal plane of the optic nerve. Cytoskeleton proteins and COX mitochondria are found in highest concentrations in the prelaminar and lamina cribrosa regions. COX and neurofilament light occurs predominantly in unmyelinated nerve, with a significant decrease in concentration occurring on optic nerve myelination. The heterogeneous distribution of cytoskeleton proteins along the sagittal plane may be an important functional adaptation that reflects the nonuniform nature of the physiological and structural environment of the optic nerve. The heterogeneous distribution of cytoskeleton proteins may also partly account for the asymmetric pattern of optic nerve damage after intraocular pressure elevation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 (Phos. and Non-Phos.) antibody produced in mouse, clone N52, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Anti-Neurofilament M (145 kDa) Antibody, CT, serum, Chemicon®
Sigma-Aldrich
Anti-MAP1 Antibody, clone HM-1, ascites fluid, clone HM-1, Chemicon®
Redes sociales
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
Investigación. Desarrollo. Producción.
Somos un proveedor líder para la industria de Ciencias de la Vida con soluciones y servicios para investigación, desarrollo y producción biotecnológicos, y para desarrollo y producción de tratamientos farmacéuticos
© 2021 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados.
Queda estrictamente prohibida la reproducción sin permiso de cualquiera de los materiales de la página web.