  • Search Results
  • Visualization of the nucleus and nuclear envelope in situ by SEM in tissue culture cells.
Visualization of the nucleus and nuclear envelope in situ by SEM in tissue culture cells.
Nature Protocols (2007)
Allen TD