PLC Plates for Separations from Milligrams to Grams
Our Preparative layer (PLC) plates allow separation and purification of samples varying in quantity – from milligrams to grams. Samples are typically applied as a band across the width of the glass plate and analyzed by UV detection. To isolate the substance by extraction, it is simply scraped from the PLC plate. We offer PLC plates with both unmodified and modified layers, in a variety of thicknesses (0.5 mm to 2 mm), and with or without fluorescent indicators.
Features and benefits
- Separation and purification from milligram to gram quantities
- Thicker layers for high sample loading
- Same proven silica-binder technology as our analytical TLC plates
PLC silica gel 60 | ||||||
|---|---|---|---|---|---|---|
| Product No. | Description | Layer Sorbent /Coating Material | Layer Thickness/ mm | Format/ cm | Support/ Backing | Pack size/ Content |
| 1.13894.0001 | PLC Silica gel 60, 0.5 mm | Silica gel 60 | 0,5 | 20 x 20 | glass | 20 plates |
| 1.05745.0001 | PLC silica gel 60, 2mm | Silica gel 60 | 2 | 20 x 20 | glass | 12 plates |
| 1.05744.0001 | PLC Silica gel 60 F254, 0.5 mm | Silica gel 60 F254*1 | 0,5 | 20 x 20 | glass | 20 plates |
| 1.13895.0001 | PLC Silica gel 60 F254, 1 mm | Silica gel 60 F254*1 | 1 | 20 x 20 | glass | 15 plates |
| 1.05717.0001 | PLC Silica gel 60 F254, 2 mm | Silica gel 60 F254*1 | 2 | 20 x 20 | glass | 12 plates |
| 1.05637.0001 | PLC Silica gel 60 F254+366, 2 mm | Silica gel 60F F254 +366*2 | 2 | 20 x 20 | glass | 12 plates |
| 1.05434.0001 | PLC Silica gel 60 RP-18 F254s, 1 mm | Silica RP-18 F254s*3 | 1 | 20 x 20 | glass | 15 plates |
| Z265829-1PAK | PLC Silica gel 60 F254, 1 mm | Silica gel 60 F254*1 | 1 | 20 x 20 | glass | 25 plates |
| Z513059-1PAK | PLC Silica gel 60 F254, 2 mm | Silica gel 60 F254*1 | 2 | 20 x 20 | glass | 25 plates |
| Z513040-1PAK | PLC Silica gel 60 F254, 1.5 mm | Silica gel 60 F254*1 | 1,5 | 20 x 20 | glass | 25 plates |
| Z513032-1PAK | PLC Silica gel 60 F254, 0.5 mm | Silica gel 60 F254*1 | 0,5 | 20 x 20 | glass | 25 plates |
*1 F254 Fluorescent indicator
*2 F254+366 Fluorescent indicator
*2 F254s Fluorescent indicator, acid stable
PLC aluminum oxide 60 and 150 | ||||||
|---|---|---|---|---|---|---|
| Product No. | Description | Layer Sorbent/ Coating Material | Layer Thickness/ mm | Format/ cm | Support/ Backing | Pack size/ Content |
| 1.05788.0001 | PLC Aluminum oxide 60 F254, 1..5 mm | Aluminum oxide 60 F254*1 | 1,5 | 20 x 20 | glass | 12 plates |
| 1.05726.0001 | PLC Aluminum oxide 60 F254, 1..5 mm | Aluminum oxide 150 F254*1 | 1,5 | 20 x 20 | glass | 12 plates |
*1 F254 Fluorescent indicator