Merck
HomeCell SignalingAssay Procedure for Creatininase
Assay Procedure for Creatininase
Principle
Creatininase Principle
The appearance of creatinine-picrate (orange dye based on Jaffe's reaction) is measured at 520nm by spectrophotometry.
Unit definition
One unit causes the formation of one micromole of orange dye per minute under the conditions described below.
Method
Reagents
Procedure
1. Pipette 1.0ml of the substrate solution (A) into a test tube and equilibrate at 37℃ for about 5 minutes.
2. Add 0.1ml of the enzyme solution* and mix.
3. After exactly 10 minutes at 37℃, immediately transfer an aliquot (0.1ml)(1/11 volume) of the reaction solution to
2.0ml of NaOH solution (B).
4. Add 1.0ml of picric acid solution (C) and incubate at 25℃ for 20 minutes.
5. Measure the optical density at 520nm against water (OD test).
At the same time, prepare the blank by transferring an aliquot (0.1ml) of the reaction solution into NaOH solution (2.0ml) just after the addition of the enzyme solution into ice-cold substrate solution, and carry out the same procedure as the test (procedure 4 and 5)(OD blank).
* Dissolve the enzyme preparation in ice-cold 50mM phosphate buffer, pH 7.5 and dilute to 1.8-2.4U/ml with the same buffer, immediately before assay.
Calculation
Activity can be calculated by using the following formula:
Volume activity
Weight activity (U/mg)=(U/ml)×1/C
Materials
Loading
This procedure is for informational purposes. For a current copy of our quality control procedure contact our Technical Service Department
Sign In To Continue
To continue reading please sign in or create an account.
Don't Have An Account?