[Saltar al contenido](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/ES/es)
Productos
Carrito0
ESES
Productos
ProductosAplicacionesServiciosRecursosApoyo
[Inicio de sesión / Registrarse](https://www.sigmaaldrich.com/oidc-sign-in)
[Búsqueda de pedido](https://www.sigmaaldrich.com/ES/es/order-lookup)
[Pedido rápido](https://www.sigmaaldrich.com/ES/es/quick-order)
Carrito0
[Home](https://www.sigmaaldrich.com/ES/en)[Solid Phase Extraction (SPE)](https://www.sigmaaldrich.com/ES/en/applications/analytical-chemistry/sample-preparation/solid-phase-extraction)LiChrolut Solid Phase Extraction Columns
# LiChrolut® Solid Phase Extraction Columns
__Section Overview:__
- [Introduction](https://www.sigmaaldrich.com#introduction)
- [LiChrolut® sorbents and columns](https://www.sigmaaldrich.com#sorbents)
- [Extraction Cartridge Selection Guide](https://www.sigmaaldrich.com#cartridge)
- [Steps of Solid Phase Extraction](https://www.sigmaaldrich.com#steps)
- [Related products](https://www.sigmaaldrich.com#products)
[Explore Products](https://www.sigmaaldrich.com#see-products)
## [](https://www.sigmaaldrich.com)Introduction
Extraction is a vital step in sample preparation before chromatographic analysis. It has been shown to enhance analyte concentration by a factor of 100 to 5,000, thereby significantly improving detection sensitivity. As part of selective and specific sample preparation, extraction ensures a more reliable and cost-effective chromatographic analysis. Offline and online extraction are two different approaches used in sample preparation. Offline extraction involves manual processing before transferring samples to an analytical instrument, while online extraction automates the process within the system itself.
We offer a range of high-quality products for offline as well as online extraction. For offline methods, the range includes [](https://www.sigmaaldrich.com)[EXtrelut® sorbents and columns for liquid-liquid extraction](https://www.sigmaaldrich.com/ES/en/technical-documents/technical-article/analytical-chemistry/solid-phase-extraction/extrelut-sle), as well as LiChrolut® sorbents and columns for solid-phase extraction.
## [](https://www.sigmaaldrich.com)LiChrolut® sorbents and columns
Solid phase extraction selectively isolates target analytes from complex or large sample volumes before their chromatographic analysis using HPLC, GC, or TLC. To ensure an efficient solid phase extraction, it is essential to avoid any interactions between the sample matrix and the stationary phase. Effective sample pretreatment, such as modification of the pH or ionic strength of the solution, is essential for emphasizing the differences between the chemical properties of the analyte and matrix components. This ensures that the analyte concentration is focused into a narrow zone on the stationary phase. The analytes can then be selectively eluted after a washing phase removes any adsorbed sample components.
LiChrolut® solid phase extraction tubes are specifically designed for reliable and rapid solid-phase extraction, using the principles of liquid chromatography. They achieve this through strong yet reversible interactions between the analyte and the stationary phase surface. Common interactions include hydrophobic forces (Van der Waals), polar forces (hydrogen bonding, dipole-dipole), or ion exchange mechanisms. These columns also offer a range of surface modifications to meet different extraction needs, including non-polar extraction (LiChrolut® RP-18 and RP-18 endcapped), polar extraction (LiChrolut® CN), cation exchange extraction (LiChrolut® SCX), mixed-mode extraction (LiChrolut® TSC).
These columns ensure outstanding consistency between batches by rigorously testing raw materials and thorough quality control on the finished product. Their production process is optimized, validated, and certified, to ensure high levels of purity and uniformity. The multi-stage purification techniques used with LiChrolut® materials are particularly beneficial for trace analysis since they lower the proportion of elutable components, resulting in purer extracts. Furthermore, all stationary phases are monitored using highly sensitive analytical methods, ensuring their adherence to extremely narrow tolerance limits.
### Features & Benefits
- Rapid sample preparation within minutes
- Higher recoveries without emulsion formation
- Highly precise analytical results using disposable cartridges
- Reduced solvent usage, allowing for lower material and disposal costs
- Possibility of automating the entire process
## [](https://www.sigmaaldrich.com)LiChrolut® Solid Phase Extraction Cartridge Selection Guide
The ideal extraction method is determined by the structure, solubility, polarity, and lipophilic characteristics (distribution coefficients) of an analyte. __Table 1__ will help you choose the most suitable LiChrolut® column for your specific application.
| | | | | |
|-------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------|------------------------------------------------|--------------------------------------------------------------------------------------------------------|-----------------------------------------------------------------------------|-------------------------------------------------------------------------------------------------------|
| Application | Extraction column | Typical sample matrix | Typical sample analytes | Typical elution solvent |
| Non-polar extraction | LiChrolut® RP-18 LiChrolut® RP-18e (endcapped) | Aqueous buffer solution | Aromatic ring systems, compounds with alkyl chains | Acetonitrile, methanol, ethyl acetate |
| Cation exchange extraction | LiChrolut® SCX (strong) | Methanolic/aqueous buffer with low ionic strength; 2 pH units under the pK value of the sample analyte | Cations: amines, pyrimidines | Aqueous buffer of high ionic strength (0.1 mol/l); 2 pH units over the pK value of the sample analyte |
| Mixed mode extraction | LiChrolut® TSC (Tox Screening Cation) | Body fluids\** | Cationic and neutral analytes | Chloroform-acetone, ammoniated ethyl acetate, or ammoniated methanol |
| Medium polar extraction of environmental pollutants | Florisil® | Waste/ground/drinking water, soil samples | Herbicides, pesticides, PCBs, PCPs, dioxins, phenols, nitro compounds, HCHs | n-Hexane, dichloromethane |
| \*\*These products are not intended for use as in-vitro diagnostics in terms of European Directive 98/79/EC. They are for research purposes only, for investigating in-vitro samples derived from the human body without any medical objective. | | | | |
Table 1.LiChrolut® selection guide
Sorbent type
High porosity synthetic silica gel particles
Particle size
40-63 μm
Pore size
60 Å
Pore volume
\-
Specific surface area
~ 600 m2/g
pH stability
pH 2-8
Wide spectra of chemically modified
phases
CN, RP-18e, RP-18,
SCX (strong cation exchanger),
TSC (Tox Screening Cation)
Capacity
\-
Table 2.Specifications of LiChrolut® materials
## [](https://www.sigmaaldrich.com)4 Steps of Solid Phase Extraction with LiChrolut® Extraction Unit
The LiChrolut® extraction unit ([](https://www.sigmaaldrich.com)[1.19852](https://www.sigmaaldrich.com/ES/en/product/mm/119852)) provides an efficient and a reliable solution for every step of solid-phase extraction. This transparent, vacuum-compatible glass unit can process up to 12 samples at the same time, delivering high throughput and consistent results. To ensure optimal analyte recovery, it is essential to carefully optimize each of the steps of the solid-phase extraction process.
1. Conditioning of the Sorbent: It is essential to solvate the chemically modified silica gels with an organic solvent, such as acetonitrile or methanol, before conditioning them. This conditioning involves preparing the gels for the sample milieu with water or buffer solution to ensure their accessibility for the analyte. It is essential to achieve reproducible sorption of the analyte. Any excess organic solvent can be removed using water or a buffer solution.
2. Application of the Sample: It involves using pressure or vacuum to introduce the sample solution into the conditioned extraction cartridges. The analyte concentrates in a narrow zone on the sorbent during this step, and it is ideal for the matrix components to neither flow through the cartridge nor be adsorbed onto it.
3. Washing: Using a small volume of water or buffer, further interfering matrix components are removed from the stationary phase's surface. A water-buffer solution with only a small quantity of methanol can also be used.
4. Elution of Adsorbed Analytes: In the last step of solid-phase extraction, the analyte is desorbed using an appropriate solvent and subsequently eluted. Following the concentration or dilution of the eluate, analysis can proceed without any delay. It is essential to choose a solvent that diminishes the interaction between the analyte and the sorbent, while also promoting an even distribution of the analyte within the eluent. The selection of a solvent must rely on a thorough understanding of the analyte’s structure, solubility, polarity, and lipophilic characteristics (distribution coefficients).
### Advantages of LiChrolut® Solid Phase Extraction Unit
- A front-positioned manometer regulates the vacuum, ensuring precise control throughout the operation.
- The glass vessel, lid, and accessories are made from inert and easy-to-clean materials, eliminating the risk of residue contamination in subsequent extractions.
- Standard accessories accommodate a wide range of collecting vessel sizes, from volumetric flasks to auto-sampler vials, offering flexibility in sample collection.
## [](https://www.sigmaaldrich.com)Related Products
Lo sentimos, se ha producido un error inesperado
Response not successful: Received status code 500
__Related Content__
- [HybridSPE-Phospholipid Technology](https://www.sigmaaldrich.com/ES/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/hybridspe-precipitation-technology)
- [Analysis of Drugs of Abuse in Urine](https://www.sigmaaldrich.com/ES/en/technical-documents/technical-article/analytical-chemistry/solid-phase-extraction/analysis-of-drugs-of-abuse-in-urine-after-cleanup-with-hlb)
- [Supel™ Swift HLB SPE Cartridges](https://www.sigmaaldrich.com/ES/en/technical-documents/technical-article/analytical-chemistry/solid-phase-extraction/supelswifthlb)
- [Method Optimization for LC-MS Analysis of Vitamin D Metabolite Critical Pairs in Serum](https://www.sigmaaldrich.com/ES/en/technical-documents/technical-article/protein-biology/protein-mass-spectrometry/vitamin-d-metabolite-critical-pairs-in-serum)
- [Reversed-Phase SPE Methodology](https://www.sigmaaldrich.com/ES/en/technical-documents/protocol/analytical-chemistry/solid-phase-extraction/reversedphase-methodology)
__Related Product Categories__
- [Solid Phase Extraction (SPE) and QuEChERS](https://www.sigmaaldrich.com/ES/en/products/analytical-chemistry/analytical-sample-preparation/spe-and-quechers)
- [Solvents](https://www.sigmaaldrich.com/ES/en/products/analytical-chemistry/analytical-chromatography/solvents)
- [Acids for laboratory, analysis, and regulated applications](https://www.sigmaaldrich.com/ES/en/products/chemistry-and-biochemicals/lab-chemicals/acids)
- [Caustic alkalis and bases](https://www.sigmaaldrich.com/ES/en/products/chemistry-and-biochemicals/lab-chemicals/caustic-alkalis-and-bases)
- [Salts](https://www.sigmaaldrich.com/ES/en/products/chemistry-and-biochemicals/lab-chemicals/salts)
- [Analytical Vials](https://www.sigmaaldrich.com/ES/en/products/analytical-chemistry/analytical-chromatography/analytical-vials)
- [Supported liquid extraction columns, resins and accessories](https://www.sigmaaldrich.com/ES/en/products/analytical-chemistry/analytical-sample-preparation/sle-columns-resins-and-accessories)
Arriba
__Inicie sesión para continuar.__
Iniciar sesión__¿No tiene una cuenta?__Registrar
Soporte[Atención al cliente](https://www.sigmaaldrich.com/ES/es/support/customer-support)[Contáctenos](https://www.sigmaaldrich.com/ES/es/collections/offices)[Preguntas frecuentes](https://maestro.my.site.com/knowledgeportal/s/)[Fichas de datos de seguridad (FDS)](https://www.sigmaaldrich.com/ES/es/documents-search?tab=sds)[Certificados (COA/COO)](https://www.sigmaaldrich.com/ES/es/documents-search?tab=coa)[Calidad y normativa](https://www.sigmaaldrich.com/ES/es/life-science/quality-and-regulatory-management)[Calculadoras y aplicaciones](https://www.sigmaaldrich.com/ES/es/support/calculators-and-apps)[Seminarios web](https://www.sigmaaldrich.com/ES/es/collections/webinars)
Pedidos[Búsqueda de pedidos](https://www.sigmaaldrich.com/ES/es/order-lookup)[Pedido rápido](https://www.sigmaaldrich.com/ES/es/quick-order)[Productos personalizados](https://www.sigmaaldrich.com/ES/es/services/custom-products)[Soluciones de comercio electrónico](https://www.sigmaaldrich.com/ES/es/life-science/ecommerce/ecommerce-solutions)
Empresa[Acerca de nosotros](https://www.sigmaaldrich.com/ES/es/life-science/about-us)[Responsabilidad](https://www.sigmaaldrich.com/ES/es/life-science/ssbi)[Eventos](https://www.sigmaaldrich.com/ES/es/collections/events)[Comunicados de prensa](https://www.sigmaaldrich.com/ES/es/collections/press)[Programas](https://www.sigmaaldrich.com/ES/es/life-science/partnership-programs)[Empleos](https://careers.merckgroup.com/global/en)[Oficinas](https://www.sigmaaldrich.com/ES/es/collections/offices)
Redes sociales
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
Merck
__Investigación. Desarrollo. Producción.__
Somos un proveedor líder en la industria global de las ciencias de la vida, con soluciones y servicios para la investigación, el desarrollo y la producción biotecnológicos, y el desarrollo y la producción de terapias farmacológicas.
[](https://www.sigmaaldrich.com/ES/es)
[SolucionesSigma-Aldrich®](https://www.sigmaaldrich.com/ES/es/life-science/sigma-aldrich)[SolucionesBio Reliance®](https://www.sigmaaldrich.com/ES/es/life-science/bioreliance)[Soluciones Millipore®](https://www.sigmaaldrich.com/ES/es/life-science/millipore)[SolucionesSAFC®](https://www.sigmaaldrich.com/ES/es/life-science/safc)[SolucionesMilli-Q®](https://www.sigmaaldrich.com/ES/es/life-science/milliq)[Soluciones Supelco®](https://www.sigmaaldrich.com/ES/es/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Alemania y/o sus filiales. Todos los derechos reservados, incluyendo la minería de textos y datos para el entrenamiento de IA y tecnologías similares.
Queda terminantemente prohibida la reproducción de cualquier material del sitio sin permiso.
[Condiciones de uso del sitio web](https://www.sigmaaldrich.com/ES/es/life-science/legal/site-use-terms)|[Política de privacidad](https://www.sigmaaldrich.com/ES/es/life-science/legal/privacy-statement)|[Condiciones generales de venta](https://www.sigmaaldrich.com/ES/es/life-science/legal/terms-and-conditions)|[Consentimiento de derechos de autor](https://www.sigmaaldrich.com/ES/es/life-science/legal/copyright-consent)|[Mapa del sitio](https://www.sigmaaldrich.com/ES/es/site-map)