Iupac Name
(isocyanatomethyl)(methyl)sulfane
Inchi Code
1S/C3H5NOS/c1-6-3-4-2-5/h3H2,1H3
InChI key
ACQSAKKIGLJHQE-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-7032350
MSDS
https://www.enaminestore.com/catalog/EN300-7032350
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service