Skip to Content
Merck

LEYH9AD00007

Leyan / Shanghai Haohong Scientific Co.,Ltd

Ethyl 4-isopropylpiperazine-1-carboxylate

Sign In to View Organizational & Contract Pricing

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

ethyl 4-isopropyl-1-piperazinecarboxylate

Inchi Code

1S/C10H20N2O2/c1-4-14-10(13)12-7-5-11(6-8-12)9(2)3/h9H,4-8H2,1-3H3

InChI key

HRMZRPQIFGCXML-UHFFFAOYSA-N

storage temp.

2-8C, protect from light

Purity

98%

Country of Origin

CN

Shipping Temperature

2-8C

Product Link

https://www.leyan.com/61014-91-3.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.leyan.com/61014-91-3.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service