Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

Apoptosis is a normal process in development and morphogenesis. However, improperly regulated apoptosis can contribute to several pathological conditions including cancer, Alzheimer′s disease, spinal muscular atrophy, ischemic cardiac damage, and autoimmune syndromes. Many cells can be activated to undergo apoptosis following the interaction of selected ligands with cell surface receptors. Receptor-mediated apoptosis involves the catalysis of preexisting intracellular caspase proenzymes into activated caspases (cysteine-containing aspartate-specific proteases). A distinctive feature of caspases is the requirement of an aspartic acid residue in the substrate P1 position. The P4 residue is important in substrate recognition and specificity. For example, caspase-1 prefers a hydrophobic tyrosine residue in the P4 position, but caspase-3 prefers an anionic aspartic residue. Caspase activity can be assayed using a fluorophore (AFC). Liberation of the fluorophore from the substrate is monitored to determine caspase activity. Supplied with a data sheet.



This convenient set of caspase substrates contains the following components, each at a concentration of 1 mM in DMSO:
Ready-to-use substrates for assaying the activity of various members of the caspase-family proteases. Contains 125 µl (1 mM) each of Caspase-1 Substrate VI, Ac-YVAD-AFC (Cat. No. 688224); Caspase-2 Substrate I, Ac-VDVAD-AFC (Cat. No. 218740); Caspase-3 Substrate IV, Ac-DEVD-AFC (Cat. No. 264150); Caspase-5 Substrate II, Ac-WEHD-AFC (Cat. No. 218754); Caspase-6 Substrate III, Ac-VEID-AFC (Cat. No. 218763); Caspase-8 Substrate I, Z-IETD-AFC (Granzyme B Substrate II, Cat. No. 368059); and Caspase-9 Substrate I, Ac-LEHD-AFC (Cat. No. 218765). Supplied with a data sheet.

Warning

Toxicity: Irritant (B)

Physical form

Each substrate supplied at 1 mM in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Humke, E.W., et al. 1998. J. Biol. Chem.273, 15702.
Thornberry, N.A., and Lazebnik, Y. 1998. Science281, 1312.
Takahashi, A., et al. 1997. Exp. Cell Res.231, 123.
Thornberry, N.A., et al. 1997. J. Biol. Chem.272, 17907.
Nicholson, D.W., et al. 1995. Nature376, 37.
Thornberry, N.A., et al. 1992. Nature356, 768.
Zimmerman, M., et al. 1977. Anal. Biochem.78, 47.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service