Skip to Content
Merck

ZRB2212

Anti-CPT2 Antibody, clone 1G7 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CPT II, Carnitine O-palmitoyltransferase 2 mitochondrial, Carnitine palmitoyltransferase II, EC:2.3.1.21


Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1G7
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 73.78 kDa
observed mol wt ~70 kDa

purified by

using Protein A

species reactivity

rat, human

species reactivity (predicted by homology)

monkey, mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CPT2(1376)

General description

Carnitine O-palmitoyltransferase 2, mitochondrial (UniProt: P23786; also known as EC:2.3.1.21, Carnitine palmitoyltransferase II, CPT II) is encoded by the CPT2 (also known as CPT1) gene (Gene ID: 1376) in human. CPT2 is a mitochondrial inner membrane protein that is involved in the synthesis of acylcarnitines from accumulated acyl-CoA metabolites. It is synthesized with a transit peptide, which is subsequently cleaved off. It catalyzes the transfer of long- and medium-chain fatty acids from cytoplasm into mitochondria, where oxidation of fatty acids takes place. It reconverts acylcarnitines back into the respective acyl-CoA esters that can then undergo β-oxidation, an essential step for the mitochondrial uptake of long-chain fatty acids and their subsequent β-oxidation. It displays higher activity with medium (C8-C12) and long-chain (C14-C18) acyl-CoA esters. Deficiency of CPT2 can result in the most common inherited disorder of long-chain fatty acid oxidation affecting skeletal muscle. Mutations in CPT2 gene can result in a lethal neonatal form, a severe infantile hepato-cardio-muscular form, or a rather mild myopathic form characterized by exercise-induced myalgia, weakness, and myoglobinuria. In CPT2 deficiency, long‐chain acylcarnitines are transported across the inner mitochondrial membrane but cannot be converted to their corresponding long‐chain acyl‐CoAs. They accumulate in the mitochondrial matrix and are transported out into plasma. These elevated levels may produce cardiac arrhythmias. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonal. (Ref.: Joshi, PR., and Zierz, S. (2020). Molecules. 25(8); 1784; Violante, S., et al. (2010). Biochim. Biophys. Acta. 1802(9); 728-732; Elpeleg, ON., et al. (2001). Am. J. Med. Genet. 102(2); 183-187).
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the mitochondrial matrix domain within the N-terminal half of human Carnitine palmitoyltransferase 2 (CPT2).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat liver tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CPT2 in Rat liver tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CPT2 in lysates from HT-29 cells and Human liver and Rat liver tissues..

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CPT2 in Human kidney and Human liver tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CPT2 in HT-29 and MCF-7 cells.

Affinity Binding Assay: A representative lot of this antibody bound CPT2 peptide with a KD of 4.1 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1G7 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Carnitine palmitoyltransferase 2 (CPT2). It targets an epitope within 18 amino acids from the mitochondrial matrix domain within the N-terminal half

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.
Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service