Sign In to View Organizational & Contract Pricing.
Select a Size
Product TA9H9A9A4774 is not currently sold in your country.
Product TA9H9A9A4774 is not currently sold in your country.
Iupac Name
(E)-5-(4-propoxybenzylidene)thiazolidine-2,4-dione
Inchi Code
1S/C13H13NO3S/c1-2-7-17-10-5-3-9(4-6-10)8-11-12(15)14-13(16)18-11/h3-6,8H,2,7H2,1H3,(H,14,15,16)/b11-8+
InChI key
GBWOSXZUTXXXQF-DHZHZOJOSA-N
storage temp.
-80/-20
Purity
99.99%
Country of Origin
CN
Shipping Temperature
4
Physical Form
solid
Product Link
https://www.targetmol.com/compound/SMI-16a
MSDS
https://www.targetmol.com/compound/SMI-16a
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.