Skip to Content
Merck
Calibration standard 3000 IR turbidity 10000 NTU±200

Calibration Standard Set

Calibration standard 3000 IR turbidity 10000 NTU±200

Products

Product Category
Brand
Physical Form
Compatibility
Technique
Showing 1-6 of 6