Merck
  • Synthesis and biological evaluation of 1-amino-2-phosphonomethylcyclopropanecarboxylic acids, new group III metabotropic glutamate receptor agonists.
Synthesis and biological evaluation of 1-amino-2-phosphonomethylcyclopropanecarboxylic acids, new group III metabotropic glutamate receptor agonists.
Journal of medicinal chemistry (2007-07-03)
Pauline Sibille, Sébastien Lopez, Isabelle Brabet, Ornella Valenti, Nadia Oueslati, Florence Gaven, Cyril Goudet, Hugues-Olivier Bertrand, Jacques Neyton, Michael J Marino, Marianne Amalric, Jean-Philippe Pin, Francine C Acher
ABSTRACT
Stereoisomers of 1-amino-2-phosphonomethylcyclopropanecarboxylic acid (APCPr), conformationally restricted analogues of L-AP4 (2-amino-4-phosphonobutyric acid), have been prepared and evaluated at recombinant group III metabotropic glutamate receptors. They activate these receptors over a broad range of potencies. The most potent isomer (1S,2R)-APCPr displays a similar pharmacological profile as that of L-AP4 (EC50 0.72, 1.95, >500, 0.34 microM at mGlu4, 6, 7, 8 receptors, respectively, and no effect at group I/II mGluRs). It was characterized on native receptors located in the basal ganglia (BG) where it induced a robust and reversible inhibition of synaptic transmission. It was tested in vivo in haloperidol-induced catalepsy, a model of Parkinsonian akinesia, by direct infusion in the globus pallidus of the BG. At a dose of 0.5 nmol/microL, catalepsy was significantly antagonized. This study reveals that (1S,2R)-APCPr is a potent group III mGluR agonist and confirms that these receptors may be considered as a therapeutic target in the Parkinson's disease.
Sigma-Aldrich
L-(+)-2-Amino-4-phosphonobutyric acid, optical purity optical purity: ≥95% (HPLC, Marfey′s reagent)
Pricing and availability is not currently available.