Merck
  • Activation of p38 and changes in mitochondria accompany autophagy to premature senescence-like phenotype switch upon chronic exposure to selenite in colon fibroblasts.
Activation of p38 and changes in mitochondria accompany autophagy to premature senescence-like phenotype switch upon chronic exposure to selenite in colon fibroblasts.
Toxicology letters (2014-09-11)
Emil Rudolf, Kateřina Rezáčová, Miroslav Cervinka
ABSTRACT
Effects of chronic exposure to supranutritional sodium selenite (Se) were investigated in colonic fibroblasts. Initially, Se did not produce any gross changes in exposed cells; however, basal levels of autophagy were transiently increased and p38 activity was stimulated. From the 3rd week onwards, Se decreased cell proliferation, with corrensponding changes in cell cycle distribution. Also, in exposed cells oxidative stress and DNA damage slowly but gradually increased along with decreasing mitochondrial function and upon continued elevated activity of p38 kinase. Towards the end of the experiment, premature senescence features became more prominent in treated cells. Pharmacological inhibition as well as gene knockdown of these processes confirmed the involvement of p38 in balancing autophagy and premature senescence in cells exposed to Se and suggests that this element may in a given time frame compromise selected cell populations in digestive system.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Cysteine
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
1-Propanesulfonic acid, technical, ≥99.0% (T)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder