Merck
  • Analysis of polymyxin B sulfate by capillary zone electrophoresis with cyclodextrin as additive. Method development and validation.
Analysis of polymyxin B sulfate by capillary zone electrophoresis with cyclodextrin as additive. Method development and validation.
Journal of chromatography. A (2000-07-13)
J W Kang, A Van Schepdael, J A Orwa, E Roets, J Hoogmartens
ABSTRACT
A capillary zone electrophoresis method for analysis of polymyxin B sulfate is described. In this method, triethanolamine (TEA)-phosphate buffer at pH 2.5 was employed to reduce the adsorption of analyte onto the capillary wall. Methyl-beta-cyclodextrin (M-beta-CD) and 2-propanol (IPA) were found to be necessary for selectivity enhancement. In order to optimize the method and to control its robustness, a central composite design was performed with four parameters, i.e. concentration of M-beta-CD, TEA, IPA and buffer pH. The optimal separation conditions were as follows: capillary, 55 cm (50 microm I.D., 47 cm effective length); 130 mM TEA-phosphate buffer (pH 2.5) containing 5 mM M-beta-CD and 5% IPA; 24 kV (51 microA) applied voltage; column temperature, 20 degrees C. Further, linearity and limits of detection quantification were examined. Three commercial samples were analyzed quantitatively.
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.