Merck
HPLC Analysis of Polar and Non-Polar Analytes on Ascentis® Express C8, Column Test Mixture
HPLC Analysis of Polar and Non-Polar Analytes on Ascentis® Express C8, Column Test Mixture application for HPLC
CONDITIONS
column
Ascentis Express C8, 5 cm × 4.6 mm I.D., 2.7 μm particles (58336-U)
mobile phase
[A] water; [B] acetonitrile; (45:55, A:B)
flow rate
1.76 mL/min
column temp.
30 °C
detector
UV, 254 nm
injection
5 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany