Merck
All Photos(1)

806943

Sigma-Aldrich

Silver

Nanowires, diam. × L 30 nm (±5 nm) × 20 μm (±3 μm), 5 mg/mL (in ETH)

EC Number:
NACRES:
NA.23

description

Flash point 57.2 °F

Quality Level

form

suspension

concentration

5 mg/mL (in ETH)

diam. × L

30 nm (±5 nm) × 20 μm (±3 μm)

color

gray

density

0.7955 g/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Owing to unique physical and chemical properties the nanowires of silver find wide range of applications extending from catalysis to photovoltaics, touch screen display, biosensing etc.

Packaging

25 mL in poly bottle

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service