Merck
All Photos(1)

GF58042324

Carbon - Vitreous

foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%

Synonym(s):
Carbon - Vitreous, VC003823
Linear Formula:
C
CAS Number:
PubChem Substance ID:

assay

96.5%

form

wire

manufacturer/tradename

Goodfellow 580-423-24

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service