All Photos(1)

1.10845

SAFC

Dihydroxyacetone

pure, not for cosmetic purposes

Synonym(s):
1,3-Dihydroxidimethyl ketone, DHA, 1,3-Dihydroxidimethyl ketone, Dihydroxyacetone
CAS Number:
Molecular Weight:
90.07
EC Index Number:
202-494-5

vapor pressure

<0.01 Pa ( 20 °C)

form

powder

potency

>16000 mg/kg LD50, oral (Rat)

pH

4-6 (20 °C, 50 g/L in H2O)

bp

188 °C (decomposition,OECD Test Guideline 103)

mp

96.5 °C (Monomer)

solubility

930 g/L

density

1.52 g/cm3 at 20 °C (OECD Test Guideline 109)

application(s)

pharmaceutical

storage temp.

2-8°C

InChI

1S/C3H6O3/c4-1-3(6)2-5/h4-5H,1-2H2

InChI key

RXKJFZQQPQGTFL-UHFFFAOYSA-N

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Dihydroxyacetone pure is a chemical intermediate.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service