Skip to Content
Merck

Skip To

1.73636

Move DC Nxt Hand-held Colorimeter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116116
NACRES:
SE.11
eCl@ss:
40050204
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Spectroquant®

technique(s)

photometry: suitable

application(s)

disinfection control

compatibility

for use with Spectroquant® test kits

storage temp.

15-25°C

General description

Spectroquant®
The Spectroquant® Move DC Nxt mobile colorimeter is suitable for the detection of five essential parameters of water analysis: chlorine, ozone, chlorine dioxide, cyanuric acid and pH. This handy, battery-operated instrument comes with a carrying case and is designed for in-process and field work.


It accelerates disinfection control workflows with rapid analysis, enhanced memory and a user-friendly Dot Matrix UI with icons and animation. The device offers a language-free UI, clear status indicators (green/red backlight), wired data transfer, differentiated measurements and a built-in Favourite list for quick method access.

Move DC Nxt Data Transfer Software (Photometer Data Transfer Tool)

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters