Skip to Content
Merck

PHR8442

Varenicline tartrate

pkg of 100 mg, certified reference material, pharmaceutical secondary standard

Synonym(s):

Varenicline tartrate, 7,8,9,10-Tetrahydro-6,10-methano-6H-pyrazino[2,3-h][3]benzazepine tartrate; (1R,12S)-5,8,14-triazatetracyclo[10.3.1.02,11.04,9]hexadeca-2,4,6,8,10-pentaene, 7,8,9,10-Tetrahydro-6,10-methano-6H-pyrazino[2,3-h][3]benzazepine tartrate, Champix

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C13H13N3 · C4H6O6
CAS Number:
Molecular Weight:
361.35
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

TWYFGYXQSYOKLK-CYUSMAIQSA-N

InChI

1S/C13H13N3.C4H6O6/c1-2-16-13-5-11-9-3-8(6-14-7-9)10(11)4-12(13)15-1;5-1(3(7)8)2(6)4(9)10/h1-2,4-5,8-9,14H,3,6-7H2;1-2,5-6H,(H,7,8)(H,9,10)/t8-,9+;1-,2-/m.1/s1

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.C1NC[C@H]2C[C@@H]1c3cc4nccnc4cc23

grade

certified reference material, pharmaceutical secondary standard

agency

(Not traceable to any primary pharmacopeia standard)

API family

varenicline

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

application(s)

pharmaceutical

format

neat

storage temp.

-10 to -25°C

Quality Level

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Biochem/physiol Actions

Varenicline tartrate is a partial α4β2 nicotinic receptor agonist and α7 full agonist.
Varenicline tartrate is a partial α4β2 nicotinic receptor agonist and α7 full agonist. Varenicline competitively binds to α4β2 receptors and partially stimulates without creating a full nicotine effect, while simultaneoudly blocking the ability of nicotine to bind to the receptors. Varenicline thus blocks the ability of nicotine to activate α4β2 receptors and stimulate the central nervous mesolimbic dopamine system, believed to be the neuronal mechanism underlying reinforcement and reward experienced upon smoking.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service