Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Level
usage
sufficient for 100 purifications
sustainability
Greener Alternative Product
concentration
45-55 mg/mL (Magnetic particle)
test parameters
total test time: 5 min
suitability
suitable for nucleic acid purification
greener alternative category
storage temp.
2-30°C
General description
Features and Benefits
- Nucleic Acid Isolation
- Manual or high-throughput automated isolations
- RT-qPCR
- Digital PCR
- Biological samples can be used
- Room temperature stable reagents
- DNA free
- DNase free
- RNase free
- NICKase free
- simple and fast protocols
Legal Information
Kit Components Only
- UPMPPART-2ML-KC Ultra pure MagPrep Magnetic Particles 1
- UPMPVRBB-40ML-KC Ultra pure MagPrep Viral RNA Lysis and Binding Buffer 1
- UPMPWB1-60ML-KC Ultra pure MagPrep Wash Buffer 1 1
- UPMPWB2-60ML-KC Ultra pure MagPrep Wash Buffer 2 1
- UPMPEB-10ML-KC Ultra pure MagPrep Elution Buffer 1
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3
target_organs
Central nervous system, Thyroid
supp_hazards
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Explore high-quality raw materials for diagnostic assay development and IVD manufacturing. Leverage our expertise in molecular diagnostic testing and enzymes for molecular assays.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| UPMPVRNA-KT | 04065272386418 |



