Skip to Content
Merck

UPMPWB1

Ultra pure MagPrep® Wash Buffer 1

greener alternative

No proteinase K or heating step needed for Magprep® Nucleic Acid Isolation,Nuclease free (NICKase, RNase, DNase), DNA-free, High-throughput Compatible and Manual

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41105500

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

usage

300-830 purifications

sustainability

Greener Alternative Product

concentration

45-55 mg/mL (Magnetic particle)

test parameters

total test time: 5 min

suitability

suitable for nucleic acid purification

greener alternative category

storage temp.

2-30°C

Quality Level

General description

The Ultra pure MagPrep® Wash Buffer 1 is intended for use within the Ultra pure MagPrep® nucleic acid purification system for manual or high throughput viral nucleic acid isolation from most biological fluids. The Ultra pure MagPrep® nucleic acid purification system is optimized for rapid easy handling and maximal yield. Nucleic acid isolation is achieved without the need for Proteinase K or sample heating, and all items are shipped and stored at ambient temperature. Buffer is quality tested to be free of contaminant nucleases and DNA, reducing the likelihood of false positives or negatives in downstream assays. Nucleic acid purified using the Ultra pure MagPrep® system is free of PCR inhibitors such as chaotropic salts or alcohols, allowing for effective amplification in various downstream applications. For research or further manufacturing use only. Not intended for direct use in humans or animals. Non-TSCA use only in US.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.

Features and Benefits

  • Nucleic Acid Isolation
  • Manual or high-throughput automated isolations
  • RT-qPCR
  • Digital PCR
  • Biological samples can be used
  • Room temperature stable reagents
  • DNA free
  • DNase free
  • RNase free
  • NICKase free
  • simple and fast protocols

Legal Information

MAGPREP is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

70.3 °F

flash_point_c

21.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore high-quality raw materials for diagnostic assay development and IVD manufacturing. Leverage our expertise in molecular diagnostic testing and enzymes for molecular assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service