Merck
All Photos(1)

52091-026

Sigma-Aldrich

DNAstable® Plus

greener alternative

10 mL bottle, for room temp preservation of DNA

NACRES:
NA.52

Quality Level

grade

for molecular biology

form

liquid

usage

400 reactions

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative category

Aligned

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

DNAstable Plus enables you to preserve, store and ship your purified DNA samples at room temperature in liquid and liquid-to-dry formats. It offers flexibility with thermostability at extreme temperatures, and the ability to handle high concentrations of DNA. Recovery from the dry matrix is achieved within minutes by simply adding water. Your DNA sample is ready for downstream analysis without the need for any additional purification steps.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product was Designed for Energy Efficiency. To see how Stanford incorporated Biomatrica Technology and saved substantial amounts of energy, see their published study:

Biomatrica technology enables safe storage of biological material at room temperature.

Packaging

1 bottle

Physical form

(1) 10 mL DNAstable Plus liquid reagent for stabilization of DNA in liquid and Liquid-to-Dry (LD) format. Also enclosed is a quick reference protocol.

Legal Information

DNAstable is a registered trademark of Biomatrica

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service