Merck
All Photos(1)

Documents

BR701452

BRAND® Reagent Reservoirs for multichannel pipettes and automatic systems

12 columns, low profile, column capacity 6 mL, pyramid bottom

Sign Into View Organizational & Contract Pricing

Synonym(s):
ldpe bottle, polyethylene bottles, polyethylene containers
NACRES:
NB.01

material

colorless
polypropylene
pyramid bottom

sterility

non-sterile

feature

RNase and DNase free
12 columns
low profile

packaging

pack of 10

manufacturer/tradename

BRAND 701452

capacity

72 mL

column capacity

6 mL

Looking for similar products? Visit Product Comparison Guide

General description

These reagent reservoirs were designed especially for use with automated liquid handling platforms. They can also be used with manual systems. Different partitions, bottom styles and profiles provide great application flexibility.

Features and Benefits

  • ANSI/SLAS standard format Compatible with nearly all liquid handling robots and other automated dispensing systems
  • 5 different types for use with multichannel pipettes and automatic systems
  • Excellent stability and chemical resistance made from high-quality polypropylene
  • Very low dead volume due to pyramid bottoms
  • Free from RNases and DNases
  • Autoclavable (121 °C, 20 min)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service