Skip to Content
Merck

91008

EPA Method 537.1 Mix

(in 1 mM NaOH in MeOH), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

certified reference material

product line

TraceCERT®

composition

N-Ethylperfluorooctanesulfonamidoacetic acid, 2000 μg/L , Heptadecafluorooctanesulfonic acid potassium salt, 2000 μg/L , N-Methylperfluorooctanesulfonamidoacetic acid, 2000 μg/L , Pentadecafluorooctanoic acid ammonium salt, 2000 μg/L , Perfluorodecanoic acid, 2000 μg/L , Perfluoroheptanoic acid, 2000 μg/L , Perfluorohexanoic acid, 2000 μg/L , Perfluorononanoic acid, 2000 μg/L , Perfluoro-2-propoxypropanoic acid, 2000 μg/L , Perfluorotetradecanoic acid, 2000 μg/L , Perfluorotridecanoic acid, 2000 μg/L , Perfluoroundecanoic acid, 2000 μg/L , Potassium 2-[(6-chloro-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexyl)oxy]-1,1,2,2-tetrafluoroethanesulfonate, 2000 μg/L , Potassium 2-[(8-chloro-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctyl)oxy]-1,1,2,2-tetrafluoroethanesulfonate, 2000 μg/L , Potassium nonafluoro-1-butanesulfonate, 2000 μg/L , Potassium perfluorohexanesulfonate, 2000 μg/L , Sodium dodecafluoro-3H-4,8-dioxanonanoate, 2000 μg/L , Tricosafluorododecanoic acid, 2000 μg/L

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

(in 1 mM NaOH in MeOH)

technique(s)

LC/MS: suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Quality Level

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Ethylperfluorooctanesulfonamidoacetic acid (EPFOS)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service