Merck
Clarification Prefilters
Examples of clarification prefilters used in pharma and biopharma production
Clarigard® and Polygard® clarification prefilters remove particulates at different steps in biopharma production. These polypropylene filters are offered in both capsule and cartridge formats, with multiple sizes to fit different application needs.
  • Clarigard® filters have a 99.99% particle retention rating, protect downstream operations, and offer low extractables and broad chemical compatibility.
  • Polygard® filters provide high dirt-holding capacity with minimal differential pressure. These nominally rated filters can be used for particulate removal from liquids and gases in multiple applications. 
Products
Sort by Relevance
Showing 1-20 of 512
Change View
Showing 1-20 of 512
Compare
Product Number
Product Name
Product Description
Pricing
Related Product Resources
Page 1 of 2