FR
EN
Applications
Products
Services
Support
Home
Search Results
Ammonia solution 32%
(1)
Ammonia solution 32%
Synonyms:
Ammonia aqueous, Ammonium hydroxide solution, Ammonia water
Linear Formula:
NH
4
OH
CAS Number:
1336-21-6
Molecular Weight:
35.05
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1.05426
Ammonia solution 32%, EMPLURA
®
Expand
View Pricing