Merck
Dodecyl sulfate sodium salt for synthesis

Dodecyl sulfate sodium salt

Dodecyl sulfate sodium salt for synthesis
Synonyms:
Dodecyl sulfate sodium salt
Empirical Formula (Hill Notation):
C12H25NaO4S
CAS Number:
Molecular Weight:
288.38

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Dodecyl sulfate sodium salt, for synthesis
Dodecyl sulfate sodium salt, for biochemistry and surfactant tests