FR
EN
Applications
Products
Services
Support
Home
Search Results
Iodide Iodate solution
(1)
Iodide Iodate solution
CAS Number:
153507-24-5
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1.60337
Iodide Iodate solution, c(I
2
) = 1/128 mol; standardised against Sodium thiosulfate solution, Titripur
®
Expand
View Pricing