
Lactophenol blue solution

Synonym(s):
Lactophenol Anilin Blue solution, Lactophenol Cotton Blue solution
Products
Shipping
Product Category
Brand
Physical Form
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing