Merck
Mitomycin C from Streptomyces caespitosus ≥970 μg/mg (USP XXIV)

Mitomycin C from Streptomyces caespitosus

Mitomycin C from Streptomyces caespitosus ≥970 μg/mg (USP XXIV)
Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture
Mitomycin C from Streptomyces caespitosus, ≥970 μg/mg (USP XXIV)