Merck
ortho-Phosphoric acid 85% Suprapur®

ortho-Phosphoric acid 85%

ortho-Phosphoric acid 85% Suprapur®
Synonyms:
Phosphoric acid, Orthophosphoric acid
Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
ortho-Phosphoric acid 85%, for analysis EMSURE® ACS,ISO,Reag. Ph Eur
ortho-Phosphoric acid 85%, for HPLC LiChropur
ortho-Phosphoric acid 85%, Suprapur®