Products
FR
EN
Products
Applications
Products
Services
Support
Home
Search Results
ortho-Phosphoric acid 99%
(1)
ortho-Phosphoric acid 99%
Synonyms:
Phosphoric acid, Orthophosphoric acid
Linear Formula:
H
3
PO
4
CAS Number:
7664-38-2
Molecular Weight:
98.00
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1.00565
ortho-Phosphoric acid 99%, cryst. for analysis EMSURE
®
Expand
View Pricing