FR
EN
Applications
Products
Services
Support
Home
Search Results
Two antioxidative lactobacilli strains as promising probiotics
Two antioxidative lactobacilli strains as promising probiotics
International Journal of Food Microbiology (2002)
Kullisaar T, et al.