  • Search Results
  • Tideglusib attenuates growth of neuroblastoma cancer stem/progenitor cells in vitro and in vivo by specifically targeting GSK-3β.
Tideglusib attenuates growth of neuroblastoma cancer stem/progenitor cells in vitro and in vivo by specifically targeting GSK-3β.
Pharmacological reports : PR (2020-10-09)
Hisham F Bahmad, Reda M Chalhoub, Hayat Harati, Jolie Bou-Gharios, Sahar Assi, Farah Ballout, Alissar Monzer, Hiba Msheik, Tarek Araji, Mohamad K Elajami, Paola Ghanem, Farah Chamaa, Humam Kadara, Tamara Abou-Antoun, Georges Daoud, Youssef Fares, Wassim Abou-Kheir
ABSTRACT
Neuroblastoma (NB) is the most frequently diagnosed extracranial solid tumor among the pediatric population. It is an embryonic tumor with high relapse rates pertaining to the presence of dormant slowly dividing cancer stem cells (CSC) within the tumor bulk that are responsible for therapy resistance. Therefore, there is a dire need to develop new therapeutic approaches that specifically target NB CSCs. Glycogen synthase kinase (GSK)-3β is a serine/threonine kinase that represents a common signaling node at the intersection of many pathways implicated in NB CSCs. GSK-3β sustains the survival and maintenance of CSCs and renders them insensitive to chemotherapeutic agents and radiation. In our study, we aimed at evaluating the potential anti-tumor effect of Tideglusib (TDG), an irreversible GSK-3β inhibitor drug, on three human NB cell lines, SK-N-SH, SH-SY5Y, and IMR-32. Our results showed that TDG significantly reduced cell proliferation, viability, and migration of the NB cells, in a dose- and time-dependent manner, and also significantly hindered the neurospheres formation eradicating the self-renewal ability of highly resistant CSCs. Besides, TDG potently reduced CD133 cancer stem cell marker expression in both SH-SY5Y cells and G1 spheres. Lastly, TDG inhibited NB tumor growth and progression in vivo. Collectively, we concluded that TDG could serve as an effective treatment capable of targeting the NB CSCs and hence overcoming therapy resistance. Yet, future studies are warranted to further investigate its potential role in NB and decipher the subcellular and molecular mechanisms underlying this role.
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA • 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture