Merck
BNP signaling is crucial for embryonic stem cell proliferation.
PloS one (2009-04-29)
Essam Mohamed Abdelalim, Ikuo Tooyama
ABSTRACT
Embryonic stem (ES) cells have unlimited proliferation potential, and can differentiate into several cell types, which represent ideal sources for cell-based therapy. This high-level proliferative ability is attributed to an unusual type of cell cycle. The Signaling pathways that regulate the proliferation of ES cells are of great interest. In this study, we show that murine ES cells specifically express brain natriuretic peptide (BNP), and its signaling is essential for ES cell proliferation. We found that BNP and its receptor (NPR-A, natriuretic peptide receptor-A) were highly expressed in self-renewing murine ES cells, whereas the levels were markedly reduced after ES cell differentiation by the withdrawal of LIF. Targeting of BNP with short interfering RNA (siRNA) resulted in the inhibition of ES cell proliferation, as indicated by a marked reduction in the cell number and colony size, a significant reduction in DNA synthesis, and decreased numbers of cells in S phase. BNP knockdown in ES cells led to the up-regulation of gamma-aminobutyric acid receptor A (GABA(A)R) genes, and activation of phosphorylated histone (gamma-H2AX), which negatively affects ES cell proliferation. In addition, knockdown of BNP increased the rate of apoptosis and reduced the expression of the transcription factor Ets-1. Appropriate BNP expression is essential for the maintenance of ES cell propagation. These findings establish BNP as a novel endogenous regulator of ES cell proliferation.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
SAFC
DMEM/Nutrient Mixture F-12 Ham, With 15 mM HEPES, without L-glutamine, L-leucine, L-lysine, L-methionine, CaCl2, MgCl2, MgSO4, sodium bicarbonate, and phenol red, powder, suitable for cell culture
Sigma-Aldrich
StableCell DMEM/F12, With stable glutamine, 15mM HEPES and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and trace elements, without HEPES and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and sodium bicarbonate, without HEPES, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture