The toxicity of three insecticides to Lysiphlebus fabarum (Marshall) (Hymenoptera: Braconidae: Aphidiinae), a parasitoid of Aphis fabae Scopoli (Hemiptera: Aphididae), was investigated using IOBC/wprs protocols. Abamectin 1.8 EC, imidacloprid 350 SC, and pymetrozine 25 WP were tested under laboratory conditions at recommended field rates. Immature stages of the parasitoid were exposed to materials by briefly dipping mummified aphids into insecticide solutions/suspensions or water (controls). Abamectin, imidacloprid, and pymetrozine caused 44.8, 58.5, and 14.5% mortality of mummies, respectively. Insecticides were also applied to broad bean foliage until run-off using a hand sprayer and the contact toxicity of residues was investigated after 1, 5, 16 and 30 day periods of outdoor weathering by caging adult wasps on treated plants for 24 h. One day-old residues of abamectin, imidacloprid, and pymetrozine produced 52.5, 90.0 and 57.0% mortality, respectively, and 5 day-old residues produced 28.1, 77.0 and 18.6% mortality. Sixteen day-old residues produced 8.8, 22.4 and 13.6% mortality, whereas 30 day-old residues produced 0.0, 3.2 and 1.1% mortality, respectively. On the basis of these results, abamectin and pymetrozine were classified as short-lived compounds (Class A) and imidacloprid as a slightly persistent compound (Class B).