  • Search Results
  • Bioprocess design for the microbial production of natural phenolic compounds by Debaryomyces hansenii.
Bioprocess design for the microbial production of natural phenolic compounds by Debaryomyces hansenii.
Applied biochemistry and biotechnology (2012-10-19)
Belén Max, Francisco Tugores, Sandra Cortés-Diéguez, José M Domínguez
ABSTRACT
Debaryomyces hansenii NRRL Y-7426 metabolised ferulic acid into different phenolic compounds using a factorial design where glucose concentration (in the range of 1-20 g/L), peptone concentration (2-20 g/L) and yeast extract concentration (0.2-10 g/L) were the independent variables. The interrelationship between dependent and operational variables was well fitted (R (2) > 0.95) to models including linear, interaction and quadratic terms. Depending on the glucose and nitrogen concentrations, which redirected the metabolism, the major degradation products were 1,226.2 mg 4-vinyl guaiacol/L after 72 h (molar yield of 86.0 %), 1,077.8 mg vanillic acid/L after 360 h (molar yield of 91.1 %) or 1,682.6 mg acetovanillone/L after 408 h (molar yield of 98.8 %) in fermentations carried out with 2,000 mg ferulic acid/L. Other metabolites such as vanillin, vanillyl alcohol or 4-ethylguaiacol were present in lower amounts.
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Supelco
Vanillic acid, certified reference material, TraceCERT®
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Methoxy-4-vinylphenol, ≥98%, FG