  • Search Results
  • Purification and characterization of a thermostable soluble peroxidase from Citrus medica leaf.
Purification and characterization of a thermostable soluble peroxidase from Citrus medica leaf.
Preparative biochemistry & biotechnology (2013-01-11)
Ruckminee Mall, Gaurav Naik, Usha Mina, Sarad Kumar Mishra
ABSTRACT
A soluble and thermostable peroxidase enzyme (POD) was extracted from the leaf of Citrus medica. The enzyme was purified 15.10-fold with a total yield of 28.6% by ammonium sulfate precipitation followed by Sephadex G-100 gel filtration chromatography. The purified enzyme came as a single band on native polyacrylamide gel electrophoresis (PAGE) as well as sodium dodecyl sulfate (SDS) PAGE. The molecular mass of the enzyme was about 32 kD as determined by SDS-PAGE. The enzyme was optimally active at pH 6.0 and 50°C temperature. The enzyme was active in wide range of pH (5.0-8.0) and temperature (30-80°C). From the thermal inactivation studies in the range of 60-75°C, the half-life (t(1/2)) values of the enzyme ranged from 8 to 173 min. The inactivation energy (Ea) value of POD was estimated to be 21.7 kcal mol(-1). The Km values for guaiacol and H(2)O(2) were 8 mM and 1.8 mM, respectively. This enzyme was activated by some metals and reagents such as Ca(2+), Cu(2+), Mg(2+), Co(2+), ferulic acid, and indole acetic acid (IAA), while it was inhibited by Fe(2+), Zn(2+), Hg(2+), and Mn(2+), L-cysteine, L-proline, and protocatechuic acid.
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material