  • Search Results
  • Stability of extemporaneous pediatric oral liquids compounded from tablets and drug substance: case of propranolol and theophylline.
Stability of extemporaneous pediatric oral liquids compounded from tablets and drug substance: case of propranolol and theophylline.
Acta poloniae pharmaceutica (2013-04-25)
Monika Muśko, Małgorzata Sznitowska
ABSTRACT
The stability of theophylline (T) and propranolol hydrochloride (P) in extemporaneously compounded oral suspensions (25 mg/mL or 50 mg/mL for T and 2 mg/mL or 5 mg/mL for P) were studied. Suspension with P and T were prepared with bulk substance or tablets using three different suspending vehicles: Ora-Sweet (M1), modified Ora-Sweet (M2) and simple syrup with glycerol and sorbitol (M3). Each suspension was stored for 35 days in a dark place at 25 degrees C and 4 degrees C. The results demonstrated that the prepared suspensions with P either from tablets or from a substance were stable in all three studied vehicles (more than 95% of initial concentration remaining). However, it is recommended that storage at 4 degrees C of suspensions prepared with M2 should be avoided because of crystallization of the buffer substances. Extemporaneous suspensions with T in an appropriate pediatric concentrations of the drug were not obtained because the problem of fast crystallization of T was not eliminated.
Product Description
Supelco
ASTM® D6584 Glycerin solution, 500 μg/mL in pyridine, analytical standard
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Theophylline, European Pharmacopoeia (EP) Reference Standard
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Theophylline solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98%
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%