  • Search Results
  • The effect of folate supplementation on methotrexate efficacy and toxicity in psoriasis patients and folic acid use by dermatologists in the USA.
The effect of folate supplementation on methotrexate efficacy and toxicity in psoriasis patients and folic acid use by dermatologists in the USA.
American journal of clinical dermatology (2013-04-12)
Amir Al-Dabagh, Scott A Davis, Megan A Kinney, Karen Huang, Steven R Feldman
ABSTRACT
Methotrexate (MTX) is an effective treatment for psoriasis but its use is limited by its toxicity. Folate supplementation can be used to reduce the adverse effects of MTX, though this may impact efficacy. The frequency of folic acid supplementation is not well characterized. The objective of this study was to review the literature involving the use of folate in patients (in particular those with psoriasis) treated with MTX and analyze trends in folic acid use. We searched PubMed from 1 May 1989 through 1 April 2012 using the terms 'folic acid,' 'folinic acid,' 'folate,' 'supplementation,' and 'methotrexate.' We also used the National Ambulatory Medical Care Survey (NAMCS) database to collect data regarding trends in MTX use and folic acid supplementation by physicians in the USA from 1993 through 2009. We assessed data including the number of MTX visits, rate of folic acid use, diagnoses, physician specialty, and demographics of patients. We used linear regression to analyze the change in folic acid use over time. Twenty-six published trials were included addressing folic acid supplementation with MTX. The majority found a benefit to folic acid supplementation, but there were only seven studies in psoriasis. Dermatologists were among the highest prescribers of MTX, and psoriasis was commonly treated with MTX. Folic acid supplementation significantly increased over this time period (p < 0.0001). However, dermatologists ranked lowest for their folate use, co-prescribing folate to only 9.1 % of MTX-treated patients. In contrast to rheumatoid arthritis, there is a scarcity of literature describing the effect of folate on MTX toxicity and efficacy in psoriasis patients. NAMCS data only included outpatient visits to non-federally employed physicians, and there is the possibility of healthcare providers not documenting over-the-counter folic acid usage. Lastly, doses of MTX and folic acid were not recorded in the database. Dermatologists were the least likely specialists to supplement MTX with folic acid. The evidence for supplementation of folic acid is mixed. The literature confirms a reduction in the adverse effects of MTX but less strongly that there may be a reduction in efficacy too. Keeping in mind the potential for folate to reduce MTX efficacy, folic acid supplementation should be considered in MTX-treated patients.
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Folic acid, European Pharmacopoeia (EP) Reference Standard
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard