  • Search Results
  • Stenotrophomonas maltophilia and Vermamoeba vermiformis relationships: bacterial multiplication and protection in amoebal-derived structures.
Stenotrophomonas maltophilia and Vermamoeba vermiformis relationships: bacterial multiplication and protection in amoebal-derived structures.
Research in microbiology (2014-12-03)
Estelle Cateau, Elodie Maisonneuve, Samuel Peguilhan, Nathalie Quellard, Yann Hechard, Marie-Helene Rodier
ABSTRACT
Stenotrophomonas maltophilia, a bacteria involved in healthcare-associated infections, can be found in hospital water systems. Other microorganisms, such as Free Living amoebae (FLA), are also at times recovered in the same environment. Amongst these protozoa, many authors have reported the presence of Vermamoeba vermiformis. We show here that this amoeba enhances S. maltophilia growth and harbors the bacteria in amoebal-derived structures after 28 days in harsh conditions. These results highlight the fact that particular attention should be paid to the presence of FLA in hospital water systems, because of their potential implication in survival and growth of pathogenic bacterial species.
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Folic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%