  • Search Results
  • Effect of epigallocatechin gallate on drug transport mediated by the proton-coupled folate transporter.
Effect of epigallocatechin gallate on drug transport mediated by the proton-coupled folate transporter.
Drug metabolism and pharmacokinetics (2014-04-04)
Maika Kissei, Tomoo Itoh, Tomoya Narawa
ABSTRACT
Folic acid (FA) is a water-soluble vitamin, and orally ingested FA is absorbed from the small intestine by the proton-coupled folate transporter (PCFT). In the present study, we investigated whether epigallocatechin gallate (EGCG), one of the tea catechins, affects the transport of FA by PCFT. EGCG inhibited the uptake of FA into Caco-2 cells and human PCFT-expressing HEK293 cells (PCFT-HEK293 cells). The initial rate of uptake of FA into PCFT-HEK293 cells followed Michaelis-Menten kinetics (K(m) = 1.9 µM). Dixon plots revealed that PCFT-mediated FA uptake was competitively inhibited by EGCG (K(i) ≒ 9 µM). The uptake of the PCFT substrate methotrexate (MTX) was competitively inhibited by EGCG as well (K(i) ≒ 15 µM). In conclusion, it is suggested that when FA or MTX is ingested with tea, it is likely that the intestinal absorption of these compounds by PCFT is inhibited, which could result in insufficient efficacy.
Product Description
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Epigallocatechin gallate, primary reference standard
Supelco
(−)-Epigallocatechin gallate, analytical standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard
Folic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Supelco
Folic acid, analytical standard
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Folic acid, meets USP testing specifications
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture